384-well high content screening microplates with glass bottom

Leverancier: Corning
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4581 4583 4585
732-2494EA 410.03 EUR
732-2494 BDAA4583 BDAA4585
384-well high content screening microplates with glass bottom
Microplaten Celcultuurplaten
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal crosstalk provide the highest possible optical quality for cell-based assays
Order Now

Specificaties

Environmentally Preferable Lage impact op de fabricage
Milieu certificering RECs, ISO 14001 - Kennebunk

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR