Peripheral IV catheters, BD Venflon™ Pro Safety
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
393226 393228 393222 393224
BDAM393226EA 191 EUR
BDAM393226 BDAM393228 BDAM393222 BDAM393224
Peripheral IV catheters, BD Venflon™ Pro Safety
Canules
These IV cannulas with polyurethane catheter and injection port are designed to access the peripheral veins of the patient’s blood system for re-hydration, parenteral nutrition, medication delivery, blood transfusion and monitoring purposes.
  • For single use
  • Needle tip protection device is activated when the steel needle is withdrawn
  • Protection device covers the steel needle tip and protects against accidental needle stick
  • Flash Back Function: During the insertion of the cannula, blood flow is visualised by blood flash back in the hub of the cannula