CytoFLEX platform flow cytometers
Leverancier: Beckman Coulter
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B53010
B53011
B53012
B53016
B53000
B53001
B53002
B53003
B53004
B53005
B53006
B53007
B53008
B53009
B53013
B53014
B53018
B53037
B96622
C02944
B75408
B75811
B75812
B78557
B78558
B78559
B78560
C02945
C02946
B78561
B96618
B96619
B96620
B96621
C02948
C00445
C00446
C02949
C11183
C11184
C01158
C01159
C01160
C01161
C02947
C11185
C11186
BECMB53010EA
0
EUR
BECMB53010
BECMB53011
BECMB53012
BECMB53016
BECMB53000
BECMB53001
BECMB53002
BECMB53003
BECMB53004
BECMB53005
BECMB53006
BECMB53007
BECMB53008
BECMB53009
BECMB53013
BECMB53014
BECMB53018
BECMB53037
BECMB96622
BECMC02944
BECMB75408
BECMB75811
BECMB75812
BECMB78557
BECMB78558
BECMB78559
BECMB78560
BECMC02945
BECMC02946
BECMB78561
BECMB96618
BECMB96619
BECMB96620
BECMB96621
BECMC02948
BECMC00445
BECMC00446
BECMC02949
BECMC11183
BECMC11184
BECMC01158
BECMC01159
BECMC01160
BECMC01161
BECMC02947
BECMC11185
BECMC11186
CytoFLEX platform flow cytometers
Celtellers
Flow Cytometers
Specificaties
Model | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX LX | CytoFLEX LX | CytoFLEX/CytoFLEX S | CytoFLEX LX | CytoFLEX LX | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX/CytoFLEX S | CytoFLEX LX | CytoFLEX LX |
Environmentally Preferable | |||||||||||||||||||||||||||||||||||||||||||||||
Gevoeligheid | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser | FITC: <30 molecules of equivalent soluble fluorochrome (MESFFITC) from the 488 nm laser; PE: <10 molecules of equivalent soluble fluorochrome (MESFPE) from the 488 nm laser |
Resolutie | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles | 488 nm Side Scatter Resolution <300 nm; 405 nm Side Scatter Resolution <80 nm polystyrene particles |
Bedrijfstemperatuur | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C | 15...27 °C |
Spanning | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V | 100 - 240 V |
b×d×h | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 605×733×451 mm | 605×733×451 mm | 425×425×340 mm | 605×733×451 mm | 605×733×451 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 425×425×340 mm | 605×733×451 mm | 605×733×451 mm |
Gewicht | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 79,0 kg | 79,0 kg | 23,4 kg | 79,0 kg | 79,0 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 23,4 kg | 79,0 kg | 79,0 kg |