Aspirator bottles
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
QL9002607
BEHRQL9002607EA 0 EUR
BEHRQL9002607
Aspirator bottles
Flessen Doseerflessen
Aspirator bottles, HDPE natural, narrow-neck storage bottles with scale.
  • UV protection for long life
  • Resistant to acids and chemicals
  • Special colour and printing possible
Storage vessel with drain made of food-safe polyethylene.