Liquid-in-glass thermometer, angled, H-B DURAC®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B608102900
BELAB608102900EA 85 EUR
BELAB608102900
Liquid-in-glass thermometer, angled, H-B DURAC®, SP Bel-Art
Thermometers Analoge thermometers
Designed for use in all heating blocks, this total immersion thermometer features a 90° angle and an arrow at 37 °C.
  • Red organic liquid against white back glass