Wash bottles, wide neck, with coloured caps, labelled, Safety-Vented™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F116420624 F116420622 F116420623 F116420638 F116430238
BELAF116420624EA 80.2 EUR
BELAF116420624 BELAF116420622 BELAF116420623 BELAF116420638 BELAF116430238
Wash bottles, wide neck, with coloured caps, labelled, Safety-Vented™, SP Bel-Art
Flessen Spuitflessen
LDPE, with PP spray cap
  • Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labeled wash bottles
  • Allows dispensing with bottle upright or inverted
  • Volatiles are vented to eliminate solvent drips
  • Pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment
Labeled with six of the most commonly used solvent names or label as per the need.More Product Information