Gas dispersion tube, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F136910000
BELAF136910000EA 145 EUR
BELAF136910000
Gas dispersion tube, SP Bel-Art
Gasbuisjes
HDPE gas dispersion tube. Bell-mouthed fitting with a sealed-in, coarse porosity Fritware® PE disc at the large end.
  • Smaller bubbles
  • Tube internal diameter: 6,35 mm
Suitable for insertion into small openings.