Funnels, thistle
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F147510000
BELAF147510000EA 143 EUR
BELAF147510000
Funnels, thistle
Trechters Filtertrechters Filtertrechters
PP, translucent
  • Chemical resistant
  • Unbreakable and safe
  • Steam autoclavable at 121 °C