Magnetic stirring bars, PTFE, cross-shaped, Spinplus®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F371440112
BELAF371440112EA 41.3 EUR
BELAF371440112
Magnetic stirring bars, PTFE, cross-shaped, Spinplus®, SP Bel-Art
Roerders Magnetische roerstaven
Spinplus® magnetic stirring bars improve speed and efficiency of mixing.
  • The '+' shape creates a deep vortex and provides stable, quiet operation
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
Certificaten: Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification