Magnetic stirring bars, PTFE, polygonal, assorted, Spinbox™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F371840000
BELAF371840000EA 311 EUR
BELAF371840000
Magnetic stirring bars, PTFE, polygonal, assorted, Spinbox™, SP Bel-Art
Roerders Magnetische roerstaven
Magnetic stirring bars with Alnico magnet, PTFE encapsulated is supplied as an assortment and comes in a convenient storage box to provide instant availability.
  • Low friction PTFE coating offers superior chemical compatibility
Leveringsinformatie: Assortment consists of 18 assorted bars, each two of the following sizes: 10×6 mm, 15×6 mm, 20×7 mm, 25×8 mm, 30×8 mm, 40×8 mm, 50×8 mm, 60×8 mm and 80×10 mm.