Desiccator cabinets, auto, vertical, secador®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F420731220 F420721220 F420741220 F420741228 F420721228
BELAF420731220EA 2520 EUR
BELAF420731220 BELAF420721220 BELAF420741220 BELAF420741228 BELAF420721228
Desiccator cabinets, auto, vertical, secador®, SP Bel-Art
Exsiccatoren Exsiccatorkasten
Secador® model 2.0, 3.0 and 4.0 auto desiccator cabinets are made from polycarbonate, which features a UV light blocker and is resistant to staining, crazing, and chemical attack.
  • Large door with easy-to-view digital hygrometer
  • Perforated shelves facilitate circulation of dry air
  • Colour-indicating silica gel allows quick visual confirmation of desiccant status
  • Amber colour units reduce visible light penetration by over 50%; perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds