Sample spoon with spatula end, Ellipso-Spoon®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H368080030 H368060015 H368060018 H368070021 H368090040
BELAH368080030EA 165 EUR
BELAH368080030 BELAH368060015 BELAH368060018 BELAH368070021 BELAH368090040
Sample spoon with spatula end, Ellipso-Spoon®, SP Bel-Art
Lepels
Stainless steel 304. The Ellipso-Spoon® sampler has a specially curved shape that permits contact with all corners of bottles and containers preventing material loss. Crystals, powders, granulated and solid substances and thick liquids are easily retrieved.
  • Autoclavable
  • Offset, deep spoon end retrieves material from the corners of bottles and containers
  • Spatula end scrapes and breaks up caked powders
  • Sampled materials never fall back into the container