Mortars without pestles, SP Bel-Art

Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H372600000 H372600001
BELAH372600000EA 916 EUR
BELAH372600000 BELAH372600001
Mortars without pestles, SP Bel-Art
Vijzels en stampers Vijzels
Liquid nitrogen cooled mortars without pestles, for grinding frozen samples.

  • Insulated handle is removable to save space; 10,2cm long
  • Three suction cup feet stabilise the unit on the workbench
  • Recommended size pestle is any size appropriate for a 275ml mortar (not included)

Grind frozen samples without liquid nitrogen hazards. This 304 stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds liquid nitrogen. The bowl is nested in a heavily insulated blue polyethylene housing.
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR