Racks for Leighton tissue culture tubes
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1917-00016
BELC1917-00016EA 374.34 EUR
BELC1917-00016
Racks for Leighton tissue culture tubes
Rekken Tube Racks
Stainless steel.
  • Spring clip prevents tube from moving in rack
  • Excellent chemical resistance
  • Excellent temperature resistance
For Leighton type culture tubes.
Order Now
Specificaties
Environmentally Preferable