Fernbach culture flasks, with DeLong® style neck
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2550-02800 2554-02800 2554-01800 2554-02833
BELC2550-02800EA 476.85 EUR
BELC2550-02800 BELC2554-02800 BELC2554-01800 BELC2554-02833
Fernbach culture flasks, with DeLong® style neck
Kolven Cultuurflessen
Manufactured from type 1 class A borosilicate glass, these flasks are designed for culturing organisms requiring a large surface area-to-volume ratio.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes