Media bottles, graduated, with screw cap
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5636-00125 5636-00250 5636-00533 5636-38500
BELC5636-00125EA 73.12 EUR
BELC5636-00125 BELC5636-00250 BELC5636-00533 BELC5636-38500
Media bottles, graduated, with screw cap
Flessen Mediumflessen
Manufactured from type 1, class B borosilicate glass.
  • White graduations
  • Marking spot
Supplied with a 33-430 nontoxic, rubber-lined black phenolic cap Large opening and sloping shoulders facilitate pouring and cleaning.