SatPax® Polx® Pre-wetted, 100% Polyester Non-woven Cleanroom Wipes, Canister and Canister Refills
Leverancier: BERKSHIRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SPXCPNW00412
BERKSPXCPNW00412EA 653 EUR
BERKSPXCPNW00412
SatPax® Polx® Pre-wetted, 100% Polyester Non-woven Cleanroom Wipes, Canister and Canister Refills
Schoonmaakdoekjes Cleanroom doekjes Vochtige cleanroom doekjes
SatPax® Polx® non-woven combines Polx non-woven wipers, composed of 100% polyester spunlace, with various saturation levels of isopropyl alcohol and DI water in an easy-to-dispense canister.
  • Exceptional cleanliness
  • Prevents abrasion with an extremely soft and pliable surface
  • Increases cleaning protocol consistency
  • Lot to lot traceability
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
In addition to being a cost-effective and easy-to-use solution, this prewetted format of Polx® non-woven has critically low fiber andMore Product Information