Recirculatie afzuigkasten, Chemcap™ Clearview™
Leverancier: BIGNEAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BC0603-4 BC1206-4 BC1506-4
BIGNBC0603-4EA 8690 EUR
BIGNBC0603-4 BIGNBC1206-4 BIGNBC1506-4
Recirculatie afzuigkasten, Chemcap™ Clearview™
Zuurkasten en behuizingen Afzuigkappen zonder afvoer
Chemcap™ Clearview™ ductless fume hoods include Chemcap™ carbon filtration. A range of six standard cabinet sizes with keypad control panel, touch button controls and service due indicator. Includes 18 W fluorescent lamp and PVC base tray which offers excellent chemical resistance and contains spillages.
Cabinets are manufactured from polyester coated steel; transparent panels are acrylic and resistant to corrosion.
  • Volledig draagbaar
  • Clearview zicht rondom
  • BC 4002 biedt envelopvormige toegang over de volledige breedte
  • BC 6003 tot BC 1204 bieden
More Product Information