Anti-Aflatoxin Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB108027-1
BIRBORB108027-1EA 0 EUR
BIRBORB108027-1
Anti-Aflatoxin Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: Aflatoxin (aflatoxin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR