Anti-Sphingosine 1 Phosphate 4 Receptor Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB108773-100
BIRBORB108773-100EA 0 EUR
BIRBORB108773-100
Anti-Sphingosine 1 Phosphate 4 Receptor Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Sphingosine 1 Phosphate 4 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR