Anti-SST Rabbit Polyclonal Antibody (Cy7®)

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB123879-100
BIRBORB123879-100EA 0 EUR
BIRBORB123879-100
Anti-SST Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Type: Primary
Antigen: SST (somatostatin)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR