Anti-ETV1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB126920-100 ORB126920-200
BIRBORB126920-100EA 0 EUR
BIRBORB126920-100 BIRBORB126920-200
Anti-ETV1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ETV1 (ets variant 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR