Anti-Monkey Milk Proteins Goat Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB22156-1
BIRBORB22156-1EA 0 EUR
BIRBORB22156-1
Anti-Monkey Milk Proteins Goat Polyclonal Antibody
Antilichamen
Type: Secondary
Antigen: Monkey Milk Proteins
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR