Anti-MX1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB41663-100
BIRBORB41663-100EA 0 EUR
BIRBORB41663-100
Anti-MX1 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MX1 (myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR