Anti-HIV protease Mouse monoclonal antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB43577-0.1
BIRBORB43577-0.1EA 0 EUR
BIRBORB43577-0.1
Anti-HIV protease Mouse monoclonal antibody
Antilichamen
Type: Primary
Antigen: HIV protease
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR