Anti-CDC37 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB67522-100
BIRBORB67522-100EA 0 EUR
BIRBORB67522-100
Anti-CDC37 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CDC37 (cell division cycle 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR