Anti-NACC1 Mouse Monoclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB69349-100
BIRBORB69349-100EA 0 EUR
BIRBORB69349-100
Anti-NACC1 Mouse Monoclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: NACC1 (nucleus accumbens associated 1, BEN and BTB (POZ) domain containing)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool NACC1
Type antilichaam Primair,nucleus accumbens associated 1, BEN and BTB (POZ) domain containing
Environmentally Preferable
Type antilichaam Primair,nucleus accumbens associated 1, BEN and BTB (POZ) domain containing
Clonality Monoklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunogeen Purified recombinant fragment of human NACC1 expressed in E. Coli.
Formaat ascites fluid
Purification purified by affinity chromatography
Concentratie 1 mg/ml
Opslagbuffer 0,03% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR