Anti-CCR5 Goat polyclonal antibody unconjugated

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB88164-50
BIRBORB88164-50EA 0 EUR
BIRBORB88164-50
Anti-CCR5 Goat polyclonal antibody unconjugated
Antilichamen
ELISA: 1:100000; Immunofluorescence: 1:300; Immunochemistry: 5 ?g/ml; Western Blot: 1:1000

Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR