Anti-CAPS2 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB89164-50
BIRBORB89164-50EA 0 EUR
BIRBORB89164-50
Anti-CAPS2 Rabbit Polyclonal Antibody
Antilichamen
Immunochemistry: 5 ?g/ml; Western Blot: 5-1 ?g/ml

Type: Primary
Antigen: CAPS2 (calcyphosine 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR