Anti-FOXA2 Mouse monoclonal antibody unconjugated

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB94663-50
BIRBORB94663-50EA 0 EUR
BIRBORB94663-50
Anti-FOXA2 Mouse monoclonal antibody unconjugated
Antilichamen
Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: FOXA2 (forkhead box A2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR