L(-)-Tyrosine

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

08125-1 08125-2
BMOL08125-1EA 0 EUR
BMOL08125-1 BMOL08125-2
L(-)-Tyrosine
L(-)-Tyrosine
Formule: C₉H₁₁NO₃
Kookpunt: 351 °C (1013 hPa)
Smeltpunt: 285 °C
Dichtheid: 1,41 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002606
CAS nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR