L(+)-Ornithine hydrochloride

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

17190-1 17190-2
BMOL17190-1EA 0 EUR
BMOL17190-1 BMOL17190-2
L(+)-Ornithine hydrochloride
L(+)-Ornithine hydrochloride
Formule: C₅H₁₂N₂O₂·HCl
MDL Nummer: MFCD00064562
CAS nummer: 3184-13-2
EINECS: 221-678-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR