D-(-)-Sorbitol

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

50204-1 50204-2
BMOL50204-1EA 0 EUR
BMOL50204-1 BMOL50204-2
D-(-)-Sorbitol
D-(-)-Sorbitol
Formule: HOH₂C(CH(OH))₄CH₂OH
Kookpunt: 295 °C (1013 hPa)
Smeltpunt: 94…98 °C
Dichtheid: 1,524 g/cm³ (20 °C)
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004708
CAS nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR