Agarose

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

50399-1 50399-2 50399-3
BMOL50399-1EA 0 EUR
BMOL50399-1 BMOL50399-2 BMOL50399-3
Agarose
Agarose
Smeltpunt: 60…90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081294
CAS nummer: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR