Decyl-β-D-glucopyranoside

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

51458-1 51458-2
BMOL51458-1EA 0 EUR
BMOL51458-1 BMOL51458-2
Decyl-β-D-glucopyranoside
Decyl-β-D-glucopyranoside
Formule: C₁₆H₃₂O₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063297
CAS nummer: 58846-77-8
EINECS: 261-469-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR