n-Nonyl-β-D-maltoside

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

52691-1 52691-2
BMOL52691-1EA 0 EUR
BMOL52691-1 BMOL52691-2
n-Nonyl-β-D-maltoside
n-Nonyl-β-D-maltoside
Formule: C₂₁H₄₀O₁₁
MDL Nummer: MFCD08062400
CAS nummer: 106402-05-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR