Natriumdodecylsulfaat 200 g/l in aqueous solution

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

54661-1 54661-2 54661-3
BMOL54661-1EA 0 EUR
BMOL54661-1 BMOL54661-2 BMOL54661-3
Natriumdodecylsulfaat 200 g/l in aqueous solution
Natriumdodecylsulfaat
Formule: CH₃(CH₂)₁₁OSO₃Na
Dichtheid: 1…1,1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036175
CAS nummer: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR