Anti-HS1 Mouse Monoclonal Antibody [clone: 3A3]

Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

KAM-TK010E
BMOLKAM-TK010EEA 0 EUR
BMOLKAM-TK010E
Anti-HS1 Mouse Monoclonal Antibody [clone: 3A3]
Antilichamen
Type: Primary
Antigen: HS1
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR