Anti-CCND1 Mouse Monoclonal Antibody (CF405S) [clone: DCS-6]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BNC040007-500 BNC040007-100
BNC040007-500EA 560 EUR
BNC040007-500 BNC040007-100
Anti-CCND1 Mouse Monoclonal Antibody (CF405S) [clone: DCS-6]
Antilichamen
Recognizes a protein of 36 kDa, identified as cyclin D1. Cyclin D1, one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. This antibody neutralizes the activity of cyclin D1 in vivo. About 60% of mantle cell lymphomas (MCL) contain a t(11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1. This antibody is useful in identifying mantle cell lymphomas (cyclin D1 positive) from CLL/SLL and follicular lymphomas (cyclin D1 negative). About 40% of breast carcinomas are positive for Cyclin D1. Occasionally, hairyMore Product Information
Order Now

Specificaties

antigeen symbool CCND1
Type antilichaam Primair,Cyclin D1
Environmentally Preferable
Type antilichaam Primair,Cyclin D1
Clonality Monoklonaal
Conjugatie CF® 405S
Clone Clone DCS-6
Reactivity Monkey,Human,Rat,Mouse
Host Mouse
Gen-ID 523852 , 667996
Isotype IgG2a kappa
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR