Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: HMPV]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNC680954-500 BNC680954-100
BNC680954-500EA 560 EUR
BNC680954-500 BNC680954-100
Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: HMPV]
Antilichamen
This MAb recognizes full-length MUC1 in a glycosylation-independent manner and can bind to the fully glycosylated protein. The dominant epitope of this MAb is APDTR in the VNTR region. It reacts with the core peptide of the MUC1 protein, which is a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (>200 kDa) and an amino acid composition rich in serine, threonine, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats that functions as multiple epitopes for theMore Product Information