Anti-CD46 Mouse Monoclonal Antibody (CF594) [clone: 197-2B1]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNC940931-500 BNC940931-100
BNC940931-500EA 560 EUR
BNC940931-500 BNC940931-100
Anti-CD46 Mouse Monoclonal Antibody (CF594) [clone: 197-2B1]
Antilichamen
Recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenicMore Product Information