Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD469]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BNUM0469-50
BNUM0469-50EA 417 EUR
BNUM0469-50
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD469]
Antilichamen
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD469
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Specificaties

Type antilichaam Primair,BrdU
Environmentally Preferable
Antigeen synoniemen BUdr|Bromodeoxyuridine
Type antilichaam Primair,BrdU
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone Clone BRD469
Reactivity Multiple
Host Mouse
Gen-ID 512843
Isotype IgG1
ImmunoChemie Yes
ImmunoFluorescentie Yes
Purification BSA-free
Concentratie 1 mg/mL
Opslagbuffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR