Anti-CYCS Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CYCS/1010]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNUM1010-50
BNUM1010-50EA 417 EUR
BNUM1010-50
Anti-CYCS Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CYCS/1010]
Antilichamen
Cytochrome C is a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of cytochrome c,More Product Information