Glass cutter for tubes
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
12210 12211
BOCH12210EA 30.5 EUR
BOCH12210 BOCH12211
Glass cutter for tubes
Snijders
Brass, nickel-plated with hard-metal wheel.
  • For glass tubes up to 40 mm in diameter
  • Replaceable steel wheel
  • Easy handling