Klemtang, precisie
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1600 1601 1860 1861
BOCH1600EA 14.4 EUR
BOCH1600 232-0006 BOCH1860 BOCH1861
Klemtang, precisie
Pincetten
These non serrated precision forceps are available in pure stainless steel or with PTFE coating.
Order Now
Specificaties
Environmentally Preferable