Drievoudige klemmen
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17021 17022 17023 17024
BOCH17021EA 1400 EUR
BOCH17021 BOCH17022 BOCH17023 BOCH17024
Drievoudige klemmen
Klemmen Klemmen voor algemeen gebruik
Holder for Schott laboratory flasks for the alternating removal of vessel and lid.
This special quick-release fastener ensures that the two laboratory flat flanges of reaction tube and flat flange lid are connected and, in addition, that the reaction tube and/or flat flange lid are held individually (optimal flange angle 45°).