Double bossheads, swivel type
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5362
BOCH5362EA 25.8 EUR
BOCH5362
Double bossheads, swivel type
Klemmen Statiefkoppelingen
Double bossheads made of malleable cast iron, chromium plated.
  • Swivel type
  • For rods up to 16,5 mm
  • Angle: 0 to 360°
  • With thumb screw (M8)