Klemmen, micro
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5405 5415 5400
BOCH5405EA 39.3 EUR
BOCH5405 BOCH5415 BOCH5400
Klemmen, micro
Klemmen Klemmen voor algemeen gebruik
Micro clamps made of stainless steel.
  • Fingers are covered with either a layer of cork or with silicone coating to protect clamped items
  • With round or angled jaws