Bekers, laag model
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A136-14 A136-15 A136-06 A136-08 A136-04 A136-13 A136-11 A136-09 A136-05 A136-03 A136-02 A136-07
BOHLA136-14EA 393 EUR
BOHLA136-14 BOHLA136-15 BOHLA136-06 BOHLA136-08 BOHLA136-04 BOHLA136-13 BOHLA136-11 BOHLA136-09 BOHLA136-05 BOHLA136-03 BOHLA136-02 BOHLA136-07
Bekers, laag model
Bekers
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.