Bekers, laag model
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A137-09 A137-07 A137-05 A137-03 A137-02 A137-01
BOHLA137-09EA 328 EUR
BOHLA137-09 BOHLA137-07 BOHLA137-05 BOHLA137-03 BOHLA137-02 BOHLA137-01
Bekers, laag model
Bekers
PFA, translucent, with spout.
  • Space-saving stackable design
  • High thermal resistance from -200 to +250 °C
  • Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.
Certificaten: FDA conform.
Order Now
Specificaties
Environmentally Preferable